Naphtho[2,1-b]furan-2-carbohydrazide ≥95%
Supplier: Matrix Scientific
045920-500MG
045920-1G
101873-270EA
700
USD
101873-270
101873-272
Naphtho[2,1-b]furan-2-carbohydrazide ≥95%
Naphtho[2,1-b]furan-2-carbohydrazide
Hazard: Irritant
Formula:
C₁₃H₁₀N₂O₂ Melting Pt: 252…255 °C |
MDL Number:
MFCD02571397 CAS Number: 53524-88-2 |
Specification Test Results
Melting point | 252-255°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...