Methyl-3-[3,5-di(tert-butyl)-4-hydroxyphenyl]propanoate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionate, 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid methyll ester, Methyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate
045916-5G
045916-25G
101873-248EA
20
USD
101873-248
101873-250
Methyl-3-[3,5-di(tert-butyl)-4-hydroxyphenyl]propanoate ≥95%
Methyl-3-[3,5-di(tert-butyl)-4-hydroxyphenyl]propanoate
Hazard: Irritant
Formula:
C₁₈H₂₈O₃ MW: 292.42 g/mol Melting Pt: 64…65 °C |
MDL Number:
MFCD00210461 CAS Number: 6386-38-5 |
Specification Test Results
Melting point | 64-65°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...