Methyl-2-benzyl-2-cyano-3-phenylpropanoate ≥95%

Supplier: Matrix Scientific

045915-500MG 045915-1G 045915-5G
101873-242EA 181.54 USD
101873-242 101873-244 101873-246
Methyl-2-benzyl-2-cyano-3-phenylpropanoate ≥95%
Methyl-2-benzyl-2-cyano-3-phenylpropanoate

Hazard: Irritant

Formula: C₁₈H₁₇NO₂
Melting Pt: 57…60 °C
MDL Number: MFCD01314310
CAS Number: 42550-72-1

Order Now

Specification Test Results

Melting point 57-60°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR