4-Nitro-2,5-xylidine ≥95%

Supplier: Matrix Scientific

Synonyms: 2,5-Dimethyl-4-nitroaniline

045914-1G 045914-5G
101873-238EA 183.08 USD
101873-238 101873-240
4-Nitro-2,5-xylidine ≥95%
4-Nitro-2,5-xylidine

Hazard: Irritant

Formula: C₈H₁₀N₂O₂
MW: 166.1772 g/mol
Melting Pt: 134-136°C
MDL Number: MFCD00025207
CAS Number: 3460-29-5

Order Now

Specification Test Results

Melting point 134-136°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR