1-(7-Bromo-5-methoxy-1-benzofuran-2-yl)-1-ethanone ≥95%

Supplier: Matrix Scientific

045913-500MG 045913-1G 045913-5G
101873-230EA 375.38 USD
101873-230 101873-232 101873-234
1-(7-Bromo-5-methoxy-1-benzofuran-2-yl)-1-ethanone ≥95%
1-(7-Bromo-5-methoxy-1-benzofuran-2-yl)-1-ethanone

Hazard: Irritant

Formula: C₁₁H₉BrO₃
Melting Pt: 123…125 °C
MDL Number: MFCD02082020
CAS Number: 454473-82-6

Order Now

Specification Test Results

Melting point 123-125°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR