3-Bromo-2-hydroxy-5-methoxybenzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-2-hydroxy-5-methoxybenzaldehyde

045912-500MG 045912-1G 045912-5G
101873-224EA 138.46 USD
101873-224 101873-226 101873-228
3-Bromo-2-hydroxy-5-methoxybenzenecarbaldehyde ≥95%
3-Bromo-2-hydroxy-5-methoxybenzenecarbaldehyde

Hazard: Irritant

Formula: C₈H₇BrO₃
MW: 231.05 g/mol
Melting Pt: 108…110 °C
MDL Number: MFCD00091938
CAS Number: 50343-02-7

Order Now

Specification Test Results

Melting point 108-110°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR