1-(p-Toluenesulfonyl)indole-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Tosyl-1H-indole-3-carbaldehyde

045911-500MG 045911-1G 045911-5G
101873-218EA 146.15 USD
101873-218 101873-220 101873-222
1-(p-Toluenesulfonyl)indole-3-carbaldehyde ≥95%
1-(p-Toluenesulphonyl)indole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₅H₁₃NO₂S
MW: 299.3443 g/mol
Melting Pt: 136…139 °C
MDL Number: MFCD00238944
CAS Number: 50562-79-3

Order Now

Specification Test Results

Melting point 136-139°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR