Ethyl-2-cyano-3-(4-hydroxyphenyl)acrylate ≥95%

Supplier: Matrix Scientific

045898-500MG 045898-1G 045898-5G
101873-162EA 181.54 USD
101873-162 101873-164 101873-166
Ethyl-2-cyano-3-(4-hydroxyphenyl)acrylate ≥95%
Ethyl-2-cyano-3-(4-hydroxyphenyl)acrylate

Hazard: Irritant

Formula: C₁₂H₁₁NO₃
Melting Pt: 180…182 °C
MDL Number: MFCD00188426
CAS Number: 6935-44-0

Order Now

Specification Test Results

Melting point 180-182°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR