2-(Benzylamino)nicotinic acid ≥95%

Supplier: Matrix Scientific

045897-500MG 045897-1G 045897-5G
101873-156EA 278.46 USD
101873-156 101873-158 101873-160
2-(Benzylamino)nicotinic acid ≥95%
2-(Benzylamino)nicotinic acid

Hazard: Irritant

Formula: C₁₃H₁₂N₂O₂
Melting Pt: 233…234 °C
MDL Number: MFCD01568566
CAS Number: 33522-80-4

Order Now

Specification Test Results

Melting point 233-234°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR