2-[(3,4,4-Trifluoro-3-butenyl)sulfanyl]nicotinohydrazide ≥95%

Supplier: Matrix Scientific

045896-500MG 045896-1G
101873-152EA 446.15 USD
101873-152 101873-154
2-[(3,4,4-Trifluoro-3-butenyl)sulfanyl]nicotinohydrazide ≥95%
2-[(3,4,4-Trifluoro-3-butenyl)sulfanyl]nicotinohydrazide

Hazard: Irritant

Formula: C₁₀H₁₀F₃N₃OS
MDL Number: MFCD01815167
CAS Number: 478247-67-5

Order Now

Specification Test Results

Melting point 168-170°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR