6-Chloro-2,3-dihydro-4H-thiochromen-4-one oxime ≥95%

Supplier: Matrix Scientific

Synonyms: N-[(4E)-6-Chloro-3,4-dihydro-2H-1-benzothiopyran-4-ylidene]hydroxylamine

045895-500MG 045895-1G 045895-5G
101873-146EA 375.38 USD
101873-146 101873-148 101873-150
6-Chloro-2,3-dihydro-4H-thiochromen-4-one oxime ≥95%
6-Chloro-2,3-dihydro-4H-thiochromen-4-one oxime

Hazard: Irritant

Formula: C₉H₈ClNOS
MW: 213.69 g/mol
Melting Pt: 138…140 °C
MDL Number: MFCD08056679
CAS Number: 1029134-50-6

Order Now

Specification Test Results

Melting point 138-140°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR