Methyl-4-[(3,4-dichlorobenzyl)oxy]benzenecarboxylate ≥95%

Supplier: Matrix Scientific

045891-500MG 045891-1G 045891-5G
101873-130EA 278.46 USD
101873-130 101873-132 101873-134
Methyl-4-[(3,4-dichlorobenzyl)oxy]benzenecarboxylate ≥95%
Methyl-4-[(3,4-dichlorobenzyl)oxy]benzenecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₂Cl₂O₃
Melting Pt: 120…122 °C
MDL Number: MFCD01568713
CAS Number: 84404-04-6

Order Now

Specification Test Results

Melting point 120-122°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR