Methyl-4-[(4-methylbenzyl)oxy]benzenecarboxylate ≥95%

Supplier: Matrix Scientific

045890-500MG 045890-1G 045890-5G
101873-124EA 278.46 USD
101873-124 101873-126 101873-128
Methyl-4-[(4-methylbenzyl)oxy]benzenecarboxylate ≥95%
Methyl-4-[(4-methylbenzyl)oxy]benzenecarboxylate

Hazard: Irritant

Formula: C₁₆H₁₆O₃
Melting Pt: 97…99 °C
MDL Number: MFCD01568711
CAS Number: 62290-48-6

Order Now

Specification Test Results

Melting point 97-99°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR