Pyrimidine-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

045887-1G 045887-5G 045887-25G
101873-112EA 15.38 USD
101873-112 101873-114 102663-724
Pyrimidine-4-carboxylic acid ≥95%
Pyrimidine-4-carboxylic acid

Hazard: Irritant

Formula: C5H4N2O2
MW: 124.10 g/mol
Melting Pt: 225 °C (dec.)
MDL Number: MFCD00129737
CAS Number: 31462-59-6

Order Now

Specification Test Results

Melting point 225°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR