Methyl-3-aminoisonicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-aminoisonicotinate, Methyl 3-aminopyridine-4-carboxylate

045885-1G 045885-5G
101873-102EA 21.54 USD
101873-102 101873-104
Methyl-3-aminoisonicotinate ≥95%
Methyl 3-aminoisonicotinate

Hazard: Irritant

Formula: C₇H₈N₂O₂
MW: 152.20 g/mol
Melting Pt: 84-86°C
MDL Number: MFCD02082535
CAS Number: 55279-30-6

Order Now

Specification Test Results

Melting point 84-86°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR