1-(Furan-3-yl)ethanone ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Acetylfuran

045884-250MG 045884-1G
101873-098EA 158.46 USD
101873-098 101873-100
1-(Furan-3-yl)ethanone ≥95%
1-(Furan-3-yl)ethanone

Hazard: Irritant

Formula: C₆H₆O₂
MW: 110.112 g/mol
Melting Pt: 47 °C
MDL Number: MFCD01568491
CAS Number: 14313-09-8

Order Now

Specification Test Results

Melting point 47°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR