4-(4-Fluorophenyl)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 1,2-Dihydro-4-(4-fluorophenyl)-2-oxopyridine-3-carbonitrile

045880-500MG 045880-1G 045880-5G
101873-076EA 375.38 USD
101873-076 101873-078 101873-080
4-(4-Fluorophenyl)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile ≥95%
4-(4-Fluorophenyl)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile

Hazard: Irritant

Formula: C₁₂H₇FN₂O
MW: 214.2 g/mol
Melting Pt: 284…285 °C
MDL Number: MFCD04117821
CAS Number: 886360-84-5

Order Now

Specification Test Results

Melting point 284-285°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR