Ethyl-3-amino-4-(dimethylamino)thieno[2,3-b]pyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

045878-500MG 045878-1G
101873-072EA 921.54 USD
101873-072 101873-074
Ethyl-3-amino-4-(dimethylamino)thieno[2,3-b]pyridine-2-carboxylate ≥95%
Ethyl-3-amino-4-(dimethylamino)thieno[2,3-b]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₅N₃O₂S
Melting Pt: 96…98 °C
MDL Number: MFCD01105700
CAS Number: 147992-86-7

Order Now

Specification Test Results

Melting point 96-98°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR