2-Chloro-4-(dimethylamino)nicotinonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-4-(dimethylamino)pyridine-3-carbonitrile

045877-500MG 045877-1G
101873-068EA 747.69 USD
101873-068 101873-070
2-Chloro-4-(dimethylamino)nicotinonitrile ≥95%
2-Chloro-4-(dimethylamino)nicotinonitrile

Hazard: Irritant

Formula: C₈H₈ClN₃
MW: 181.63 g/mol
Melting Pt: 84…86 °C
MDL Number: MFCD00563985
CAS Number: 147992-80-1

Order Now

Specification Test Results

Melting point 84-86°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR