4-(Dimethylamino)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile ≥95%

Supplier: Matrix Scientific

045876-500MG 045876-1G 045876-5G
101873-062EA 921.54 USD
101873-062 101873-064 101873-066
4-(Dimethylamino)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile ≥95%
4-(Dimethylamino)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile

Hazard: Irritant

Formula: C₈H₉N₃O
MW: 163.18 g/mol
Melting Pt: 282…284 °C
MDL Number: MFCD01314755
CAS Number: 62321-91-9

Order Now

Specification Test Results

Melting point 282-284°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR