N-(5-Iodo-2-pyridinyl)acetamide ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Acetamido-5-iodopyridine, 2-Acetylamino-5-iodopyridine

045873-500MG 045873-1G 045873-5G
101873-044EA 278.46 USD
101873-044 101873-046 101873-048
N-(5-Iodo-2-pyridinyl)acetamide ≥95%
N-(5-Iodo-2-pyridinyl)acetamide

Hazard: Irritant

Formula: C₇H₇IN₂O
MW: 262.05 g/mol
Melting Pt: 156…158 °C
MDL Number: MFCD01315262
CAS Number: 66131-78-0

Order Now

Specification Test Results

Melting point 156-158°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR