2-[4-(3,3-Dimethyl-2-oxo-1-azetanyl)phenyl]acetonitrile ≥95%

Supplier: Matrix Scientific

045871-500MG 045871-1G 045871-5G
101873-038EA 747.69 USD
101873-038 101873-040 101873-042
2-[4-(3,3-Dimethyl-2-oxo-1-azetanyl)phenyl]acetonitrile ≥95%
2-[4-(3,3-Dimethyl-2-oxo-1-azetanyl)phenyl]acetonitrile

Hazard: Irritant

Formula: C₁₃H₁₄N₂O
MW: 214.27 g/mol
Melting Pt: 87…89 °C
MDL Number: MFCD00793454
CAS Number: 478262-00-9

Order Now

Specification Test Results

Melting point 87-89°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR