3-Chloro-N-[4-(cyanomethyl)phenyl]-2,2-dimethylpropanamide ≥95%

Supplier: Matrix Scientific

045870-500MG 045870-1G 045870-5G
101873-032EA 375.38 USD
101873-032 101873-034 101873-036
3-Chloro-N-[4-(cyanomethyl)phenyl]-2,2-dimethylpropanamide ≥95%
3-Chloro-N-[4-(cyanomethyl)phenyl]-2,2-dimethylpropanamide

Hazard: Irritant

Formula: C₁₃H₁₅ClN₂O
Melting Pt: 116…118 °C
MDL Number: MFCD00793453
CAS Number: 454473-74-6

Order Now

Specification Test Results

Melting point 116-118°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR