3-Chloro-N-(3,5-dimethoxyphenyl)-2,2-dimethylpropanamide ≥95%
Supplier: Matrix Scientific
045868-500MG
045868-1G
045868-5G
101873-020EA
181.54
USD
101873-020
101873-022
101873-024
3-Chloro-N-(3,5-dimethoxyphenyl)-2,2-dimethylpropanamide ≥95%
3-Chloro-N-(3,5-dimethoxyphenyl)-2,2-dimethylpropanamide
Hazard: Irritant
Formula:
C₁₃H₁₈ClNO₃ Melting Pt: 129…131 °C |
MDL Number:
MFCD00793443 CAS Number: 454473-71-3 |
Specification Test Results
Melting point | 129-131°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...