4-(3,3-Dimethyl-2-oxo-1-azetanyl)benzenecarbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(3,3-Dimethyl-2-oxoazetidin-1-yl)benzonitrile

045866-500MG 045866-1G 045866-5G
101873-008EA 747.69 USD
101873-008 101873-010 101873-012
4-(3,3-Dimethyl-2-oxo-1-azetanyl)benzenecarbonitrile ≥95%
4-(3,3-Dimethyl-2-oxo-1-azetanyl)benzenecarbonitrile

Hazard: Irritant

Formula: C₁₂H₁₂N₂O
MW: 200.24 g/mol
Melting Pt: 121…123 °C
MDL Number: MFCD00793440
CAS Number: 454473-69-9

Order Now

Specification Test Results

Melting point 121-123°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR