N-[3,5-Bis(trifluoromethyl)phenyl]-3-chloro-2,2-dimethylpropanamide ≥95%

Supplier: Matrix Scientific

045865-500MG 045865-1G 045865-5G
101873-002EA 181.54 USD
101873-002 101873-004 101873-006
N-[3,5-Bis(trifluoromethyl)phenyl]-3-chloro-2,2-dimethylpropanamide ≥95%
N-[3,5-Bis(trifluoromethyl)phenyl]-3-chloro-2,2-dimethylpropanamide

Hazard: Irritant

Formula: C₁₃H₁₂ClF₆NO
MW: 347.69 g/mol
Melting Pt: 119…121 °C
MDL Number: MFCD00793437
CAS Number: 478261-82-4

Order Now

Specification Test Results

Melting point 119-121°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR