2-Amino-1-benzyl-4,5-diphenyl-1H-pyrrole-3-carbonitrile ≥95%

Supplier: Matrix Scientific

045859-500MG 045859-1G 045859-5G
101872-968EA 75.38 USD
101872-968 101872-970 101872-972
2-Amino-1-benzyl-4,5-diphenyl-1H-pyrrole-3-carbonitrile ≥95%
2-Amino-1-benzyl-4,5-diphenyl-1H-pyrrole-3-carbonitrile

Hazard: Irritant

Formula: C₂₄H₁₉N₃
Melting Pt: 175…179 °C
MDL Number: MFCD01935933
CAS Number: 55817-67-9

Order Now

Specification Test Results

Melting point 175-179°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR