3-Bromo-2-(2-bromoacetyl)thiophene ≥95%
Supplier: Matrix Scientific
045854-500MG
045854-1G
045854-5G
101872-938EA
124.62
USD
101872-938
101872-940
101872-942
3-Bromo-2-(2-bromoacetyl)thiophene ≥95%
3-Bromo-2-(2-bromoacetyl)thiophene
Hazard: Irritant
Formula:
C₆H₄Br₂OS Melting Pt: 54…56 °C |
CAS Number:
81216-95-7 |
Specification Test Results
Melting point | 54-56°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...