3-(1,3-Benzothiazol-2-yl)-2-pyridinol ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(1,3-Benzothiazol-2-yl)-2-hydroxypyridine

045849-500MG 045849-1G 045849-5G
101872-914EA 278.46 USD
101872-914 101872-916 101872-918
3-(1,3-Benzothiazol-2-yl)-2-pyridinol ≥95%
3-(1,3-Benzothiazol-2-yl)-2-pyridinol

Hazard: Irritant

Formula: C₁₂H₈N₂OS
MW: 228.27 g/mol
Melting Pt: 305…306 °C
MDL Number: MFCD08443951
CAS Number: 292140-76-2

Order Now

Specification Test Results

Melting point 305-306°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR