3-Methoxy-4-[(4-methoxybenzyl)oxy]benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(4-Methoxybenzyloxy)-3-Methoxybenzaldehyde

045847-500MG 045847-1G 045847-5G
101872-908EA 278.46 USD
101872-908 101872-910 101872-912
3-Methoxy-4-[(4-methoxybenzyl)oxy]benzenecarbaldehyde ≥95%
3-Methoxy-4-[(4-methoxybenzyl)oxy]benzenecarbaldehyde

Hazard: Irritant

Formula: C₁₆H₁₆O₄
MW: 272.3 g/mol
Melting Pt: 113…115 °C
MDL Number: MFCD00202691
CAS Number: 129047-38-7

Order Now

Specification Test Results

Melting point 113-115°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR