2-Oxo-6-phenyl-1,2-dihydro-3-pyridinecarbonitrile ≥95%
Supplier: Matrix Scientific
045846-500MG
045846-1G
045846-5G
101872-902EA
278.46
USD
101872-902
101872-904
101872-906
2-Oxo-6-phenyl-1,2-dihydro-3-pyridinecarbonitrile ≥95%
2-Oxo-6-phenyl-1,2-dihydro-3-pyridinecarbonitrile
Hazard: Irritant
Formula:
C₁₂H₈N₂O MW: 196.21 g/mol Melting Pt: 353…355 °C |
MDL Number:
MFCD00202791 CAS Number: 43083-13-2 |
Specification Test Results
Melting point | 353-355°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...