1-(4-Fluorophenyl)-2-(methylsulfonyl)-1-ethanone ≥95%

Supplier: Matrix Scientific

Synonyms: 4'-Fluoro-2-(methylsulphonyl)acetophenone

045845-500MG 045845-1G 045845-5G
101872-896EA 278.46 USD
101872-896 101872-898 101872-900
1-(4-Fluorophenyl)-2-(methylsulfonyl)-1-ethanone ≥95%
1-(4-Fluorophenyl)-2-(methylsulfonyl)-1-ethanone

Hazard: Irritant

Formula: C₉H₉FO₃S
MW: 216.23 g/mol
Melting Pt: 111…113 °C
MDL Number: MFCD00202594
CAS Number: 82652-17-3

Order Now

Specification Test Results

Melting point 111-113°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR