Methyl-2-cyano-3-(dimethylamino)acrylate ≥95%

Supplier: Matrix Scientific

045844-500MG 045844-1G 045844-5G
101872-890EA 89.23 USD
101872-890 101872-892 101872-894
Methyl-2-cyano-3-(dimethylamino)acrylate ≥95%
Methyl-2-cyano-3-(dimethylamino)acrylate

Hazard: Irritant

Formula: C₇H₁₀N₂O₂
Melting Pt: 105…107 °C
MDL Number: MFCD00086506
CAS Number: 1187-27-5

Order Now

Specification Test Results

Melting point 105-107°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR