Methyl-4-hydroxy-1-[(4-methylphenyl)sulfonyl]-2-pyrrolidinecarboxylate ≥95%
Supplier: Matrix Scientific
045843-500MG
045843-1G
045843-5G
101872-884EA
747.69
USD
101872-884
101872-886
101872-888
Methyl-4-hydroxy-1-[(4-methylphenyl)sulfonyl]-2-pyrrolidinecarboxylate ≥95%
Methyl-4-hydroxy-1-[(4-methylphenyl)sulfonyl]-2-pyrrolidinecarboxylate
Hazard: Irritant
Formula:
C₁₃H₁₇NO₅S Melting Pt: 97…99 °C |
MDL Number:
MFCD00202789 CAS Number: 16257-57-1 |
Specification Test Results
Melting point | 97-99°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...