2-(Aminooxy)-N-(4-chlorobenzyl)acetamide hydrochloride ≥95%

Supplier: Matrix Scientific

045842-500MG 045842-1G 045842-5G
101872-878EA 747.69 USD
101872-878 101872-880 101872-882
2-(Aminooxy)-N-(4-chlorobenzyl)acetamide hydrochloride ≥95%
2-(Aminooxy)-N-(4-chlorobenzyl)acetamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₂Cl₂N₂O₂
Melting Pt: 173…176 °C
MDL Number: MFCD01114503
CAS Number: 324009-20-3

Order Now

Specification Test Results

Melting point 173-176°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR