2-(Aminooxy)-N-(2-methoxyphenyl)acetamide hydrochloride ≥95%

Supplier: Matrix Scientific

045839-500MG 045839-1G 045839-5G
101872-872EA 747.69 USD
101872-872 101872-874 101872-876
2-(Aminooxy)-N-(2-methoxyphenyl)acetamide hydrochloride ≥95%
2-(Aminooxy)-N-(2-methoxyphenyl)acetamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₃ClN₂O₃
Melting Pt: 172…174 °C
MDL Number: MFCD01114502
CAS Number: 34242-84-7

Order Now

Specification Test Results

Melting point 172-174°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR