4-Chloro-N'-(2-chloroacetyl)benzenecarbohydrazide ≥95%
Supplier: Matrix Scientific
045838-500MG
045838-1G
045838-5G
101872-866EA
181.54
USD
101872-866
101872-868
101872-870
4-Chloro-N'-(2-chloroacetyl)benzenecarbohydrazide ≥95%
4-Chloro-N'-(2-chloroacetyl)benzenecarbohydrazide
Hazard: Irritant
Formula:
C₉H₈Cl₂N₂O₂ Melting Pt: 183…184 °C |
MDL Number:
MFCD00793105 CAS Number: 50677-27-5 |
Specification Test Results
Melting point | 183-184°C |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...