2-(Aminooxy)-1-morpholino-1-ethanone hydrochloride ≥95%

Supplier: Matrix Scientific

045837-500MG 045837-1G 045837-5G
101872-860EA 730.77 USD
101872-860 101872-862 101872-864
2-(Aminooxy)-1-morpholino-1-ethanone hydrochloride ≥95%
2-(Aminooxy)-1-morpholino-1-ethanone hydrochloride

Hazard: Irritant

Formula: C₆H₁₃ClN₂O₃
Melting Pt: 202…204 °C
MDL Number: MFCD01114501
CAS Number: 312521-29-2

Order Now

Specification Test Results

Melting point 202-204°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR