2-(Aminooxy)-N-(2-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]amino}ethyl)acetamide hydrochloride ≥95%

Supplier: Matrix Scientific

045834-500MG 045834-1G 045834-5G
101872-854EA 1483.08 USD
101872-854 101872-856 101872-858
2-(Aminooxy)-N-(2-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]amino}ethyl)acetamide hydrochloride ≥95%
2-(Aminooxy)-N-(2-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]amino}ethyl)acetamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₃Cl₂F₃N₄O₂
Melting Pt: 157…160 °C
MDL Number: MFCD01114568
CAS Number: 324009-15-6

Order Now

Specification Test Results

Melting point 157-160°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR