Methyl-2-{[(4-chlorophenyl)sulfonyl]amino}-2-phenylacetate ≥95%

Supplier: Matrix Scientific

045833-500MG 045833-1G 045833-5G
101872-848EA 375.38 USD
101872-848 101872-850 101872-852
Methyl-2-{[(4-chlorophenyl)sulfonyl]amino}-2-phenylacetate ≥95%
Methyl-2-{[(4-chlorophenyl)sulfonyl]amino}-2-phenylacetate

Hazard: Irritant

Formula: C₁₅H₁₄ClNO₄S
MW: 339.8 g/mol
Melting Pt: 101…103 °C
MDL Number: MFCD01935995
CAS Number: 454473-65-5

Order Now

Specification Test Results

Melting point 101-103°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR