4-{[3-(Trifluoromethyl)phenyl]sulfanyl}aniline ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-3'-(trifluoromethyl)diphenyl sulphide

045832-500MG 045832-1G 045832-5G
101872-842EA 921.54 USD
101872-842 101872-844 101872-846
4-{[3-(Trifluoromethyl)phenyl]sulfanyl}aniline ≥95%
4-{[3-(Trifluoromethyl)phenyl]sulfanyl}aniline

Hazard: Irritant

Formula: C₁₃H₁₀F₃NS
MW: 269.29 g/mol
Melting Pt: 67…69 °C
MDL Number: MFCD00172345
CAS Number: 339104-66-4

Order Now

Specification Test Results

Melting point 67-69°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR