1-Chloroacetyl-4-[3-chloro-5-(trifluoromethyl)pyrid-2-yl]piperazine ≥95%

Supplier: Matrix Scientific

045829-500MG 045829-1G 045829-5G
101872-826EA 747.69 USD
101872-826 101872-828 101872-830
1-Chloroacetyl-4-[3-chloro-5-(trifluoromethyl)pyrid-2-yl]piperazine ≥95%
1-Chloroacetyl-4-[3-chloro-5-(trifluoromethyl)pyrid-2-yl]piperazine

Hazard: Irritant

Formula: C₁₂H₁₂Cl₂F₃N₃O
Melting Pt: 85…87 °C
MDL Number: MFCD00172868
CAS Number: 260553-15-9

Order Now

Specification Test Results

Melting point 85-87°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR