2-{4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]piperazino}acetohydrazide ≥95%

Supplier: Matrix Scientific

045827-500MG 045827-1G 045827-5G
101872-814EA 1109.23 USD
101872-814 101872-816 101872-818
2-{4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]piperazino}acetohydrazide ≥95%
2-{4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]piperazino}acetohydrazide

Hazard: Irritant

Formula: C₁₂H₁₅ClF₃N₅O
Melting Pt: 121…123 °C
MDL Number: MFCD00141696
CAS Number: 338979-10-5

Order Now

Specification Test Results

Melting point 121-123°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR