6-Methyl-2-(2-pyridinyl)-4-pyrimidinol ≥95%

Supplier: Matrix Scientific

045825-500MG 045825-1G 045825-5G
101872-802EA 224.62 USD
101872-802 101872-804 101872-806
6-Methyl-2-(2-pyridinyl)-4-pyrimidinol ≥95%
6-Methyl-2-(2-pyridinyl)-4-pyrimidinol

Hazard: Irritant

Formula: C₁₀H₉N₃O
MW: 187.2 g/mol
Melting Pt: 114…116 °C
MDL Number: MFCD00114146
CAS Number: 55417-80-6

Order Now

Specification Test Results

Melting point 114-116°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR