4-{[5-(Trifluoromethyl)-2-pyridinyl]oxy}aniline ≥95%

Supplier: Matrix Scientific

Synonyms: 4-((5-(Trifluoromethyl)pyridin-2-yl)oxy)aniline

045822-500MG 045822-1G 045822-5G
101872-790EA 120 USD
101872-790 101872-792 101872-794
4-{[5-(Trifluoromethyl)-2-pyridinyl]oxy}aniline ≥95%
4-{[5-(Trifluoromethyl)-2-pyridinyl]oxy}aniline

Hazard: Irritant

Formula: C₁₂H₉F₃N₂O
MW: 254.21 g/mol
Melting Pt: 44…47 °C
MDL Number: MFCD03001213
CAS Number: 71422-81-6

Order Now

Specification Test Results

Melting point 44-47°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR