4-(3-Chlorobenzoyl)-1-methyl-1H-pyrrole-2-carbaldehyde ≥95%
Supplier: Matrix Scientific
045821-500MG
045821-1G
045821-5G
101872-784EA
124.62
USD
101872-784
101872-786
101872-788
4-(3-Chlorobenzoyl)-1-methyl-1H-pyrrole-2-carbaldehyde ≥95%
4-(3-Chlorobenzoyl)-1-methyl-1H-pyrrole-2-carbaldehyde
Hazard: Irritant
Formula:
C₁₃H₁₀ClNO₂ Melting Pt: 137…139 °C |
MDL Number:
MFCD00173497 CAS Number: 338978-14-6 |
Specification Test Results
Melting point | 137-139°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...