5-Chloro-2-oxo-1-[4-(trifluoromethyl)benzyl]-1,2-dihydro-3-pyridine carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Chloro-1-[4-(trifluoromethyl)benzyl]pyridin-2-one-3-carboxylic acid

045817-500MG 045817-1G 045817-5G
101872-766EA 700 USD
101872-766 101872-768 101872-770
5-Chloro-2-oxo-1-[4-(trifluoromethyl)benzyl]-1,2-dihydro-3-pyridine carboxylic acid ≥95%
5-Chloro-2-oxo-1-[4-(trifluoromethyl)benzyl]-1,2-dihydro-3-pyridine carboxylic acid

Hazard: Irritant

Formula: C₁₄H₉ClF₃NO₃
MW: 331.68 g/mol
Melting Pt: 197…199 °C
MDL Number: MFCD00243767
CAS Number: 338977-66-5

Order Now

Specification Test Results

Melting point 197-199°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR