4-Benzoyl-1H-pyrrole-2-carboxamide ≥95%

Supplier: Matrix Scientific

045814-500MG 045814-1G 045814-5G
101872-748EA 747.69 USD
101872-748 101872-750 101872-752
4-Benzoyl-1H-pyrrole-2-carboxamide ≥95%
4-Benzoyl-1H-pyrrole-2-carboxamide

Hazard: Irritant

Formula: C₁₂H₁₀N₂O₂
Melting Pt: 230…232 °C
MDL Number: MFCD00141917
CAS Number: 338976-94-6

Order Now

Specification Test Results

Melting point 230-232°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR