5,7-Bis(trifluoromethyl)[1,8]naphthyridin-2-ol ≥95%

Supplier: Matrix Scientific

045812-500MG 045812-1G 045812-5G
101872-736EA 375.38 USD
101872-736 101872-738 101872-740
5,7-Bis(trifluoromethyl)[1,8]naphthyridin-2-ol ≥95%
5,7-Bis(trifluoromethyl)[1,8]naphthyridin-2-ol

Hazard: Irritant

Formula: C₁₀H₄F₆N₂O
Melting Pt: 185 °C
MDL Number: MFCD00140206
CAS Number: 51420-73-6

Order Now

Specification Test Results

Melting point 185°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR