4-(4-Bromophenyl)-2-oxo-2,5-dihydro-3-furancarbonitrile ≥95%

Supplier: Matrix Scientific

045809-500MG 045809-1G 045809-5G
101872-718EA 200 USD
101872-718 101872-720 101872-722
4-(4-Bromophenyl)-2-oxo-2,5-dihydro-3-furancarbonitrile ≥95%
4-(4-Bromophenyl)-2-oxo-2,5-dihydro-3-furancarbonitrile

Hazard: Irritant

Formula: C₁₁H₆BrNO₂
Melting Pt: 215…217 °C
MDL Number: MFCD00139239
CAS Number: 7721-24-6

Order Now

Specification Test Results

Melting point 215-217°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR